Gerald Dressing Forceps
Gerald Dressing Forceps
Actual product may vary
Gerald Dressing Forceps
Serrated design
Have Questions?
Catalog number 6184
Price (SGD)
217.00
EA
Add to cart
Price (SGD)
217.00
EA
Add to cart
  • Length 7 in. (17.8cm)
  • Satin-finish stainless steel
Specifications
Tip StyleCurved, Serrated
TypeChart Recorder Paper
Length (English)7 in.
MaterialStainless Steel
Temperature Range-40°C to +25°C
Length (Metric)178 mm
Product TypeForceps, Dressing, Gerald
Quantity (No. per Case)Each
FinishSatin
For Use With (Equipment)Free-Standing and Panel-Mounted Chart Recorders
Monitoring OptionsRecorder
Static Holding Time7 Days
Diameter (English)6 in.
Diameter (Metric)15.2 cm
Unit SizeEA