Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Actual product may vary
Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Have Questions?
Catalog NumberLength (Metric)
6169-0010248 mm
Catalog number 6169-0010
Price (SGD)
55.00
PK
Add to cart
Length (Metric):
248 mm
Price (SGD)
55.00
PK
Add to cart
Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.
  • Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
  • Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
  • Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications
Specifications
AutoclavableAutoclavable
ColorWhite
DescriptionStirring Rod with flat paddle and triangular paddle ends
Length (English)9¾ in.
Length (Metric)248 mm
MaterialPolypropylene
TypeStirring Rod
Unit SizePK