Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Actual product may vary
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge
Easily operate these repairable hand-operated PVC vacuum pumps, ideal for siphoning, filtration, liquid transfer, bleeding fluid lines and checking for leaks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFlow RateFor Use With
6132-002036 cc/stroke6132-3000 Repair Kit
6132-001015 cc/stroke6132-2000 Repair Kit
FKS1041199136 cc/stroke6132-3000 Repair Kit
Catalog number 6132-0020
Price (SGD)
317.00
EA
Add to cart
Flow Rate:
36 cc/stroke
For Use With:
6132-3000 Repair Kit
Price (SGD)
317.00
EA
Add to cart
Easily operate Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Vacuum Pumps with Gauge, ideal for siphoning, filtration, liquid transfer, bleeding fluid lines and checking for leaks. Lightweight units are sealed and self-lubricating with removable covers on exhaust port and trigger to release vacuum without disconnecting pumps from the line. Quickly attain and hold vacuum of 25 in. (635 mm) Hg. The vacuum trigger release requires only one hand to operate and releases vacuum with the touch of an index finger.
Features:
  • Gauge monitors vacuum in kPa and inches of mercury for visual indication of pump rate and progress.
  • New handle design is more durable and ergonomic.
  • Adjustable vacuum release rate provides control.
  • Full blank port pressure is obtained with only two strokes, making operation efficient.
  • Removable cover on exhaust port and trigger allow release vacuum without disconnecting pumps from the line.
  • Adjustable vacuum release rate is controlled by a trigger on the pump.
  • Nozzle fits standard 0.25 in. (6 mm) I.D. tubing.
  • System can be disassembled for repair and replacement of internal parts. (see catalog numbers for repair kits in the specifications tables)
Includes:
2′ (0.6 m) Nalgene 180 clear plastic tubing
.
Specifications
DescriptionNalgene repairable PVC hand pump with gauge
No. per Pack1
No. per Case4
Max. Vacuum25 in. Hg
Flow Rate36 cc/stroke
TypeVacuum Pump
For Use With6132-3000 Repair Kit
Unit SizeEA