HPLC UV Detector Lamps
HPLC UV Detector Lamps
Actual product may vary
HPLC UV Detector Lamps
HPLC UV Detector Lamps can be used with Variable Wavelength, Diode Array, and Multiple-Wavelength detectors.
Have Questions?
Catalog NumberType
6074.1110
also known as 6074.1110S
Deuterium Lamp
Catalog number 6074.1110
also known as 6074.1110S
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Deuterium Lamp
Choose Thermo Scientific™ HPLC UV Detector Lamps for use with Variable Wavelength, Diode Array, and Multiple-Wavelength detectors.
Specifications
Detection ModeUV Detection
TypeDeuterium Lamp
For Use With (Equipment)DAD-3000(RS), MWD-3000(RS) and VWD-3x00(RS) Series Detector
Unit SizeEA