1x1 image pixel for data collection
Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Actual product may vary
Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Shake multiple tubes at an adjustable angle to maximize sample aeration. The rack holders are adjustable to fit any of the four available test tube rack sizes.
Have Questions?
Catalog NumberProduct TypeTube Size
600088Test Tube Rack HolderVaries, Adjustable
Catalog number 600088
Price (SGD)
1,015.00
EA
Add to cart
Product Type:
Test Tube Rack Holder
Tube Size:
Varies, Adjustable
Price (SGD)
1,015.00
EA
Add to cart
Shake multiple tubes at an adjustable angle to maximize sample aeration. Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide ultimate flexibility—they can be adjusted to fit any of the four available test tube rack sizes.
  • Universal test tube rack holder can be adjusted to fit any of the four available test tube rack sizes
  • Universal design requires the separate purchase of a test tube rack
Specifications
Tube SizeVaries, Adjustable
Product TypeTest Tube Rack Holder
DescriptionUniversal adjustable-angle test tube rack holder
For Use With (Equipment)MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link