MSG 606, Tocris Bioscience™
MSG 606, Tocris Bioscience™
MSG 606, Tocris Bioscience™
MSG 606, Tocris Bioscience™
Thermo Scientific
MSG 606, Tocris Bioscience™
Potent human MC1 receptor antagonist; also MC3 and MC5 partial agonist
Have Questions?
Catalog NumberQuantity
595411 mg
Catalog number 59541
HSN 39269099
Price (SGD)
398.00
EA
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (SGD)
398.00
EA
Add to cart
Specifications
CAS1416983-77-1
Recommended StorageStore at −20°C
Molecular FormulaC62H82N20O13S
Linear FormulaC62H82N20O13S
Quantity1 mg
View more
Potent human MC1 receptor antagonist (IC50 = 17 nM). Also partial agonist at human MC3 and MC5 receptors (EC50 values are 59 and 1300 nM, respectively). Exhibits binding affinity for A375 melanoma cells in vitro. Reverses morphine-induced hyperalgesia in female mice, with no effect in male mice. γMSH analog.