Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Actual product may vary
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Material (Membrane)Pore SizeFits Bottle Neck Size (Metric)
595-4520500 mLPES0.2 μm45mm
596-3320150 mLPES0.2 μm33mm
FKS10441411150 mLPES0.2 μm33mm
596-4520150 mLPES0.2 μm45mm
296-4545150 mLPES0.45 μm45mm
595-3320500 mLPES0.2 μm33mm
FIS09-741-07500 mLPES0.2 μm45mm
295-4545500 mLPES0.45 μm45mm
FIS09-740-64B500 mLPES0.45 μm45mm
FKS10646421500 mLPES0.45 μm45mm
597-45201000 mLPES0.2 μm45mm
FIS09-741-111000 mLPES0.2 μm45mm
FIS09-740-22K500 mLSFCA0.2 μm45mm
291-4545500 mLSFCA0.45 μm45mm
FIS09-740-22L500 mLSFCA0.45 μm45mm
FKS101398001000 mLSFCA0.2 μm33mm
Catalog number 595-4520
HSN 84212900
Price (SGD)
135.00
Special offer
Ends: 30-Jun-2026
206.00
Save 71.00 (34%)
Case
Volume (Metric):
500 mL
Material (Membrane):
PES
Pore Size:
0.2 μm
Fits Bottle Neck Size (Metric):
45mm
Price (SGD)
135.00
Special offer
Ends: 30-Jun-2026
206.00
Save 71.00 (34%)
Case
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
Specifications
DisposableSingle-use
Filter TypeMembrane Filter
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.2 μm
SterilitySterile
Fits Bottle Neck Size (Metric)45mm
Green FeaturesACT Ecolabel (Gold)
No. per Case12
No. per Pack12
TypeBottle Top Filter
Volume (Metric)500 mL
Diameter (Metric)75 mm
Diameter (Metric) Membrane75 mm
Fit for PurposeSterile
Unit SizeCase
Frequently asked questions (FAQs)
Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?
No, the filter membranes for the bottle top filters are molded in and cannot be removed.