Nalgene™ Polycarbonate General Purpose Test Tube Racks
Nalgene™ Polycarbonate General Purpose Test Tube Racks
Actual product may vary
Nalgene™ Polycarbonate General Purpose Test Tube Racks
Nalgene™ Polycarbonate General Purpose Test Tube Racks
Nalgene™ Polycarbonate General Purpose Test Tube Racks
Thermo Scientific
Nalgene™ Polycarbonate General Purpose Test Tube Racks
Organize and store test tubes in these convenient tube racks which feature handles for easy carrying, and molded-in numbers and letters for easy tube identification.
Have Questions?
Catalog NumberArrayFor Use With (Equipment)
5929-00303 x 825-30mm Tubes
Catalog number 5929-0030
Price (SGD)
355.00
CS
Add to cart
Array:
3 x 8
For Use With (Equipment):
25-30mm Tubes
Price (SGD)
355.00
CS
Add to cart
Organize and store test tubes using Thermo Scientific™ Nalgene™ Polycarbonate General Purpose Test Tube Racks. These racks are available with handles for easy carrying. When empty, they can be stacked with any other racks of the same length. Holes are keyed with molded-in numbers and letters for easy identification of tubes.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float
  • Autoclavable (will reduce strength over time)
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Product TypeTest Tube Rack
DescriptionTest Tube Rack PC 30mm
Array3 x 8
AutoclavableAutoclavable
ColorClear
HoldsTubes (25 to 30 mm)
MaterialPolycarbonate
Positions24
ShapeRectangular
StackableYes
For Use With (Equipment)25-30mm Tubes
No. per Case4
No. per Pack1
Length (Metric)32 cm
Length (English)12.5 in.
Width (Metric)11.5 cm
Width (English)4.5 in.
Diameter (Metric) Opening30 mm
Diameter (Metric) Inner30 mm
Unit SizeCS