Check Order Status
18/10 Stainless Steel Retort Rings
Actual product may vary
18/10 Stainless Steel Retort Rings
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Retort Rings comes in three types.
Have Questions?
Catalog Number
Product Type
Length (Metric)
Diameter (Metric) Outer
5500
also known as S5500
Closed Ring
160 mm
50 mm
Catalog number
5500
also known as S5500
Price (SGD)
362.00
CS
Add to cart
Product Type:
Closed Ring
Length (Metric):
160 mm
Diameter (Metric) Outer:
50 mm
Price (SGD)
362.00
CS
Add to cart
Made from 18/10 stainless steel
Three types: Closed ring, Closed ring with bosshead, and Open ring
Specifications
Material
Stainless Steel (18/10)
Length (Metric)
160 mm
Diameter (Metric) Outer
50 mm
Product Type
Closed Ring
Quantity
1 Pk.