VIOS 160L 3 Door Inner Gas Tight Screen Door
Product Image
VIOS 160L 3 Door Inner Gas Tight Screen Door
VIOS 160L 3 Door inner gas tight screen door
Have Questions?
Catalog NumberCapacity (Metric)
51901144160 L
Catalog number 51901144
Price (SGD)
3,038.00
EA
Add to cart
Capacity (Metric):
160 L
Price (SGD)
3,038.00
EA
Add to cart
VIOS 160L 3 Door inner gas tight screen door
VIOS 160L 3 Door inner gas tight screen door
Specifications
Capacity (Metric)160 L
TypeScreen Door
Quantity3 Door
Unit SizeEA