Search Thermo Fisher Scientific
Catalog Number | Description | Type | For Use With (Equipment) |
---|---|---|---|
51900297 | Eurotherm 2404/P4 temperature controller with RS 232 interface | Eurotherm | VT 6060 M and VT 6130 M |
51900193 | Pressure controller with solenoid valve | Pressure Controller | VT 6060 M/P and VT6130 M/P |
51900195 | Digicon™ S Temperature controller, 300°C | Temperature Controller | VT 6060 P,VT 6130 P |
51900196 | Digicon™ S Temperature controller, 400°C | Temperature Controller | VT 6060 P |
51900209 | Thermicon* P temperature program controller | Thermicon | VT 6060 M,VT6130 M |
51900073 | 20 hour timer (24 hr at 50Hz) | Digital Timer | VT 6060 M/P and VT6130 M/P |
51900205 | Daily Program Timer | Program Timer | VT 6060 M/P and VT6130 M/P |
51900079 | Rated temperature of 400°C (door without inspection window) | Door | VT 6060 P |
51900076 | Vacuum chamber with rounded corners at the rear | Vacuum Chamber | VT 6130 M |
51900069 | Digital Pressure Display | Digital Pressure Display | VT 6060 M/P and VT6130 M/P |
51900081 | Central monitoring connection | Central Monitoring Connection | VT 6060 M/P and VT6130 M/P |
50028403 | Additional shelf (incl. Shelf supports) | Shelf | VT 6025 |
50043976 | Additional shelf (incl. Shelf supports) | Shelf | VT 6130 M |
50029597 | Support frame, 780mm height | Support Frame | VT 6130 M/P |
50048618 | Stainless-steel tray | Accessories | VT 6130 M/P |
50048619 | Stainless-steel tray | Tray | VT6130 M/P |