Heraguard™ ECO Clean Bench
Actual product may vary
Heraguard™ ECO Clean Bench
Heraguard™ ECO Clean Bench
Heraguard™ ECO Clean Bench
Heraguard™ ECO Clean Bench
Heraguard™ ECO Clean Bench
Protect samples with the Thermo Scientific™ Heraguard™ ECO Clean Bench, which features optimal efficiency, safety, and functionality. Available with dual DC motors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDescriptionMaterialWidth (English)
51029702ECO Clean BenchHeraguard Eco 1.2Melamine4 ft.
51029701ECO Clean BenchHeraguard Eco 0.9Melamine3 ft.
51029703ECO Clean BenchHeraguard Eco 1.5Melamine5 ft.
51029704ECO Clean BenchHeraguard Eco 1.8Melamine6 ft.
51029705ECO Clean BenchHeraguard Eco 1.8/95Melamine6 ft.
51900303Line Cord And Internal ReceptacleUKMelamine
Catalog number 51029702
HSN 84146000
Price (SGD)
5,463.50
Special offer
Ends: 30-Nov-2026
10,927.00
Save 5,463.50 (50%)
Each
Product Type:
ECO Clean Bench
Description:
Heraguard Eco 1.2
Material:
Melamine
Width (English):
4 ft.
Price (SGD)
5,463.50
Special offer
Ends: 30-Nov-2026
10,927.00
Save 5,463.50 (50%)
Each
Protect samples with the Thermo Scientific™ Heraguard™ ECO Clean Bench, which features optimal efficiency, safety and functionality. Available with DC motors for low energy consumption, the clean bench meets ISO class 5 minimum conditions achieved in the work area and EU GMP Grade A work environment. The horizontal laminar flow technology is used in a variety of applications where sample protection is required: hospitals, pharmaceutical applications, cleanrooms, and industrial applications.
Clean Benches Provide:
  • Sample protection
  • Clean air conditions that can be validated
  • Ease of use
Variety of Interior Widths:
  • 900mm
  • 1200mm
  • 1500mm
  • 1800mm
Different Working Heights:
  • Standard work space height 650mm
  • Extra work space height 950mm
All-Around Vision
  • Side windows increase the available light inside the work area and support a comfortable work environment
Compliance:
  • Agency: GOST
  • CE: 2006/95/EC; 2004/108/EC; 2011/65/EC; 2006/121/EC
  • Standards: EN 61010-1:2011; EN 61326:2006
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
Product TypeECO Clean Bench
MaterialMelamine
Air VelocityLevel I: 0.15 to 0.25 m/s, Level II: 0.36 to 0.45 m/s
Depth (English) Interior23.03 in.
Depth (Metric) Interior585 mm
DescriptionHeraguard Eco 1.2
Height (English)46.06 in.
Height (English) Interior25.39 in.
Height (Metric)1170 mm
Height (Metric) Interior645 mm
Illumination>1000 lx
Length (Metric)810 mm
Load Bearing Capacity (Metric)30 kg
ModelECO 1.2
Noise LevelLevel I: < 55, Level II: < 60 dBA
Ports3
Power Consumption270 W
Voltage230 V
Weight (Metric)120 kg
Width (English)4 ft.
Width (English) Interior48.03 in.
Width (Metric)1300 mm
Width (Metric) Interior1220 mm
Window MaterialTempered Safety Glass