Heratherm™ General Protocol Microbiological Incubators
Heratherm™ General Protocol Microbiological Incubators
Actual product may vary
Heratherm™ General Protocol Microbiological Incubators
Heratherm™ General Protocol Microbiological Incubators
Heratherm™ General Protocol Microbiological Incubators
Perform a range of general applications with these general protocol incubators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Voltage
51029334747 L230 V
5102813075 L230 V
51028131117 L230 V
FKS10839810117 L230 V
51028132194 L230 V
51029322405 L230 V
Catalog number 51029334
Price (SGD)
14,115.00
EA
Add to cart
Capacity (Metric):
747 L
Voltage:
230 V
Price (SGD)
14,115.00
EA
Add to cart
Use Thermo Scientific™ Heratherm General Protocol Microbiological Incubators for routine applications. The Heratherm General Protocol Incubator line is designed with sample safety in mind, for applications in pharmaceutical, medical, food and research laboratories.
  • Gravity convection provides gentle air flow and minimal drying out
  • Temperature range from ambient 5° to 75°C
  • Temperature uniformity of ±0.6°C (measured at 37°C)
  • Temperature stability of ±0.2°C (measured at 37 °C)
  • Corrosion-resistant stainless-steel chamber (AISI 430)
  • Intuitive user interface for setting temperature
  • Large, easy to read vacuum fluorescent display
  • Internal glass door allows sample viewing without impacting temperature
  • Exceptionally small footprint
Ordering Information:
Availability may vary by country.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)26.4 cu. ft.
Certifications/ComplianceCE marked
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)747 L
Chamber MaterialStainless Steel
Convection TechnologyGravity convection
MaterialStainless Steel
No. of Shelves2 supplied/ 39 max.
Plug TypeCountry-specific
Temperature Range (Metric)Ambient +5° to 75°C
TypeMicrobiological Incubator
Temperature Stability±0.4°C @ 37°C
Temperature Uniformity±1.3°C @ 37°C
Dimensions (D x W x H) Interior22.4 x 39.5 x 51.5 in. (56.9 x 100.4 x 130.7 cm)
Dimensions (L x W x H)30.3 x 49.7 x 60.8 in. (77 x 126.1 x 154.5 cm)
Length (English)30.3 in.
Length (Metric)77 cm
Width (English)49.7 in.
Width (English) Interior39.5 in.
Width (Metric)126.1 cm
Width (Metric) Interior100.4 cm
Height (English)60.8 in.
Height (English) Interior51.5 in.
Height (Metric)154.5 cm
Height (Metric) Interior130.7 cm
Depth (English) Interior22.4 in.
Depth (Metric) Interior56.9 cm
Weight (English)443 lb.
Weight (Metric)201 kg
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Wattage1500 w
Amperage6.5 A