Nalgene™ Microcentrifuge Tube Storage Boxes
Nalgene™ Microcentrifuge Tube Storage Boxes
Actual product may vary
Nalgene™ Microcentrifuge Tube Storage Boxes
Nalgene™ Microcentrifuge Tube Storage Boxes
Nalgene™ Microcentrifuge Tube Storage Boxes
Nalgene™ Microcentrifuge Tube Storage Boxes
Safely store samples using these microcentrifuge tube storage boxes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberArrayFor Use With (Equipment)
5055-50158 x 81.5mL tubes
FIS15-350-588 x 80.2mLtubes
FIS15-350-599 x 90.5mL tubes
FKS101499508 x 81.5mL tubes
Catalog number 5055-5015
Price (SGD)
104.00
PK
Add to cart
Array:
8 x 8
For Use With (Equipment):
1.5mL tubes
Price (SGD)
104.00
PK
Add to cart
Safely store samples using Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes. High-temperature polyurethane foam insert securely holds 64 or 81 microcentrifuge tubes sized 0.2, 0.5 or 1.5mL.
  • Bottom constructed of white polycarbonate
  • Transparent polycarbonate lid has printed grid for inventory purposes
  • Store from -211° to +250°F (-135° to +121°C)
  • Lid accepts marker for box and tube identification and is keyed to orient front and back of box
  • L x W x H: 5.25 x 5.25 x 2 in. (13 x 13 x 5cm)
Note:
Not for use in liquid phase of liquid nitrogen.
Specifications
Product TypeMicrocentrifuge Tube Storage Box
Array8 x 8
ColorWhite
HoldsMicrotubes (1.5 mL)
MaterialPolyurethane Foam, Polycarbonate
Positions64
ShapeSquare
For Use With (Equipment)1.5mL tubes
No. per Case24
No. per Pack4
Unit SizePK