Heracell VIOS Support Frame
Product Image
Heracell VIOS Support Frame
Lower profile support frame with casters
Have Questions?
Catalog NumberdimVoltage
50154551
Catalog number 50154551
Price (SGD)
1,177.00
EA
Add to cart
Price (SGD)
1,177.00
EA
Add to cart
Lower profile support frame with casters, for the 165L models of Heracell VIOS
Heracell VIOS Support frame
For the 165L models of Heracell VIOS.
Lower profile support frame with casters.
Specifications
DescriptionLower profile support frame with casters
For Use With (Equipment)165 L Models of Heracell VIOS
TypeVIOS Support Frame
Unit SizeEA