Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Actual product may vary
Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Increase flexibility with additional remote dispensers
Have Questions?
Catalog NumberTypeHeight (English)Height (Metric)
50136494Water Dispenser, Bench Version28.5 in.725 mm
Catalog number 50136494
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Water Dispenser, Bench Version
Height (English):
28.5 in.
Height (Metric):
725 mm
  • Available in bench and wall-mounted versions
  • Attach up to two additional dispensers per system
Specifications
For Use With (Equipment)GenPure xCAD Plus, GenPure xCAD Plus UV, GenPure xCAD Plus UF, GenPure xCAD Plus UV/UF, GenPure xCAD Plus UV - TOC, GenPure xCAD Plus UV/UF-TOC
TypeWater Dispenser, Bench Version
Width (English)10.2 in.
Width (Metric)260 mm
Height (English)28.5 in.
Height (Metric)725 mm
Depth (English)21 in.
Depth (Metric)530 cm
Unit SizeEA