1x1 image pixel for data collection
Barnstead™ LabTower™ RO Water Purification System
Barnstead™ LabTower™ RO Water Purification System
Actual product may vary
Barnstead™ LabTower™ RO Water Purification System
Convert tap water into reverse osmosis water with integrated 100L storage using the Thermo Scientific™ Barnstead™ LabTower™ RO Water Purification System.
Have Questions?
Catalog NumberDescriptionPermeate performanceElectrical Requirements
50132391LabTower RO 40L/hr.40 L/hr. at 10°C100/240 V 50/60 Hz
Catalog number 50132391
Price (SGD)
18,358.00
EA
Estimated availability date 09-Sep-2025
Add to cart
Description:
LabTower RO 40L/hr.
Permeate performance:
40 L/hr. at 10°C
Electrical Requirements:
100/240 V 50/60 Hz
Price (SGD)
18,358.00
EA
Add to cart
Convert tap water into reverse osmosis (RO) water with integrated 100L storage using the Thermo Scientific™ Barnstead™ LabTower™ RO Water Purification System, which allows the user to easily read parameters with an ultra-modern controller. Mounted on casters, this compact system is an ideal pure water supply for any laboratory. The system is well-suited as a feed water source for dishwashers, autoclaves and general laboratory use. Choose from two system options with permeate flows of 20, 49 or 60L/hr. Resistivity/conductivity is clearly displayed on a large back-lit control panel that tilts for optimum viewing.
Advanced technology in a mobile design:
  • An ultra-modern microprocessor control provides easy-to-read system parameters.
  • Completely drainable integrated 100L high-purity water reservoir has a low-noise pressure booster as a standard component.
  • Mounted on casters, the compact LabTower RO system is an ideal pure water supply for any laboratory.
  • System is ideally suited as a feed water source for dishwashers, autoclaves and general laboratory use.
  • A built-in pretreatment unit — consisting of a hardness stabilizer for protection of the reverse osmosis module from hardness formers and an activated carbon/5μm combi-cartridge — protects the system against free chlorine and particles, ensuring the long service life of downstream purification stages.
Compact and mobile design:
  • System includes three system options with permeate flows of 20, 40 or 60L/hr.
  • Stand-alone design with integrated 100L tank takes up ZERO bench space—casters allow for easy relocation.
  • Systems can be upgraded later to accommodate growing water capacity demands.
Integrated 100L reservoirL
  • System stores 100L of RO water in a high-purity polyethylene PE opaque reservoir.
  • Adjustable reservoir volume can be programmed for times of low demand—dispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.
Easy-to-operate clear display:
  • Resistivity/conductivity clearly displayed on large back-lit control panel that tilts for optimum viewing.
  • Status of current operating mode clearly indicating “production”, “stand-by”, “cleaning” or “disinfection” modes.
  • Reservoir fill-level is shown as a percent.
  • System parameters are code-protected to prevent accidental changes to set points.
GLP-compliant documentation:
  • LabTower RO has been developed to fulfill GLP requirements.
  • Recorded and traceable data can be obtained by printout via the RS-232 interface and accessory printer.
  • Highly qualified and precise measurement of the conductivity is ensured by the cell constant of 0.16 cm-1.
Pretreatment:  
  • External Pretreatment required
Specifications
Bacterial Content99%
DescriptionLabTower RO 40L/hr.
Feed Water MonitoringYes
Feedwater SourceTap
Flow Rate40 L/hr. at 10°C
TOC Monitor with UV Intensity MonitoringNo
TypeReverse Osmosis Water Purification System
Feedwater Connector3/4 in. NPT
Feedwater Pressure1 to 6 Bar
Operating Pressure2 to 6 bar
Particles >0.2µM/mL<1 μM/mL
Permeate performance40 L/hr. at 10°C
Power Consumption0.25 kW
Silica Removal98%
Temperature Range (Metric)2°C to 35°C
Width (Metric)450 mm
Height (Metric)1500 mm
Depth (Metric)580 mm
Net Weight (Metric)50 kg
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V
Frequency50/60 Hz
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link