Barnstead™ Smart2Pure™ Water Purification System
Actual product may vary
Barnstead™ Smart2Pure™ Water Purification System
Convert tap water to ASTM Type 1 water with this compact, all-in-one water system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelRO Flow Rate
50129845Smart2Pure 12 UV/UF12 L/hr.
50129688Smart2Pure 3 UV/UF3 L/hr.
50129872Smart2Pure 3 UV3 L/hr.
50129885
also known as 50129885PR
Smart2Pure 6 UV6 L/hr.
50129887Smart2Pure 6 UV/UF6 L/hr.
50129890Smart2Pure 12 UV12 L/hr.
Catalog number 50129845
HSN 84212190
Price (SGD)
9,087.00
Special offer
Ends: 30-Jun-2026
11,358.00
Save 2,271.00 (20%)
Each
Model:
Smart2Pure 12 UV/UF
RO Flow Rate:
12 L/hr.
Price (SGD)
9,087.00
Special offer
Ends: 30-Jun-2026
11,358.00
Save 2,271.00 (20%)
Each
Convert tap water to both ASTM Type 1 and Type 2 water with the compact, all-in-one Thermo Scientific™ Barnstead™ Smart2Pure™ Water Purification System. This complete system produces 3 L or 6 L of ultrapure water per hour. The Smart2Pure 3 and 6 features a built-in 6 L reservoir to store the Type 2 water, while the Smart2Pure 12 offers the choice of a 30 L or 60 L reservoir for customizable storage capacity. Dispensing is easy and features variable speed to control flow. The display can be tilted for optimal reading. Position this system on the bench or mount on the wall.
Lasting Economy:
  • Water purification progresses through independent cartridges with Aquastop quick connects for fast replacement.
  • Module 1 offers a combination of pretreatment and the reverse osmosis membrane.
  • Module 2 includes a polishing cartridge that contains high-quality, ultrapure resin for consistent purity and long cartridge life.
Easy-to-Operate:
  • Dispensing is easy and features variable speed to control flow.
  • The display can be tilted for optimal reading.
  • Optional UV bulb and UF ultrafilter are available to customize ultrapure water.
  • Flexible mounting on wall or bench.
Smart Integrated 6L Reservoir for the Smart2Pure 3 and 6:
  • Constructed with pigment-free materials.
  • Conical bottom outlet for complete draining and efficient cleaning and disinfection.
Smart2Pure 12 Offers Reservoir Options (30L or 60L):
  • Large opening allows easy and effective cleaning by hand.
  • Sterile venting filter/reservoir overflow protects against contamination by microorganisms.
  • Optional CO2 absorber prevents TOC value increases from drawn-in CO2.
  • Polyethylene reservoir is opaque to light.
  • Recirculation pump protects the high purity water from bacterial growth during standstills and maintains the low conductivity value.
  • Wall-mount is optional for simple space-saving.
  • Conical bottom outlet allows for complete draining and efficient cleaning and disinfection.
Optional Hand Dispenser:
  • Gain increased flexibility by adding a hand dispenser with 3m cord (Part #50138221)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Bacterial retention99%
ConductivityType 1: 0.055μS/cm
Bacterial Content<0.01 CFU/mL
Feedwater Connector3/4 in. NPT
Feed Water MonitoringNo
Feedwater SourceTap
Flow Rate0.6 L/min.
Product Water TypeType 1
ResistivityType 1: 18.2 Mohm.cm at 25°C
TOC Monitor with UV Intensity MonitoringNo
Total Organic Carbon1 to 5 ppb
Feedwater Pressure1 to 6 bar
IncludesRO/pretreatment cartridge, an ultrapure polisher cartridge, sterile 0.2μm filter, pressure regulator, UV lamp and UF filter.
ModelSmart2Pure 12 UV/UF
Operating Pressure1 to 6 bar
Particles >0.2µM/mL<1
Permeate performance12 L/hr. at 10°C
Power Consumption0.06 kW
RO Flow Rate12 L/hr.
Width (Metric)305 mm
Height (Metric)545 mm
Depth (Metric)400 mm
Net Weight (Metric)22 kg
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V
Frequency50/60 Hz
Endotoxin Concentration0.005 EU/mL
Product TypeWater Purification System
Unit SizeEach