Heratherm Wire Mesh Shelf
Product Image
Heratherm Wire Mesh Shelf
Wire mesh shelf for Heratherm 180L ovens
Have Questions?
Catalog number 50127766
Price (SGD)
452.00
EA
Add to cart
Price (SGD)
452.00
EA
Add to cart
Wire mesh shelf for Heratherm 180L ovens, Includes 2 shelf supports
Heratherm Wire mesh shelf
For Heratherm 180L ovens
Includes 2 shelf supports
Specifications
For Use With (Equipment)Heratherm 180 L Ovens
TypeOven Accessories
IncludesIncludes 2 shelf supports
Unit SizeEA