Cimarec™ 40M Controllers
Cimarec™ 40M Controllers
Actual product may vary
Cimarec™ 40M Controllers
Maintain constant speeds, even during viscosity changes, with our 40M controllers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelVoltage
50118911Telemodul 80 M Control Units (80w)230 V UK
50118904Telemodul 40 M Control Units (40w)230 V UK
Catalog number 50118911
Price (SGD)
5,099.00
EA
Add to cart
Model:
Telemodul 80 M Control Units (80w)
Voltage:
230 V UK
Price (SGD)
5,099.00
EA
Add to cart
Maintain constant speeds, even during viscosity changes, with Thermo Scientific™ Cimarec™ 40M Controllers. This product is required for use with the Thermo Scientific™ Cimarec™ Mobil Direct.
Features:
  • Adjustable speeds from 100 to 1000rpm
  • High visibility digital display
  • Adjustable start-up times
  • Thermal and electrical overload protection, with fault alert
Telemodul 40 M
  • Powerful controller equips Mobil 200 and 600 stirrers for the most demanding mixing tasks
Telemodul 80 M
  • Offers all the functionality of the Telemodul 40 M, with power output boosted to 80W for more demanding stirring tasks
Warranty:5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ModelTelemodul 80 M Control Units (80w)
Speed100 to 1000 rpm
TypeStirrer Controller
Voltage230 V UK
Electrical Requirements230 V
Stirring Range100 RPM to 1000 RPM
Unit SizeEA