Cimarec™ Mono & Maxi Direct Stirrers
Cimarec™ Mono & Maxi Direct Stirrers
Actual product may vary
Cimarec™ Mono & Maxi Direct Stirrers
Cimarec™ Mono & Maxi Direct Stirrers
Cimarec™ Mono & Maxi Direct Stirrers
Cimarec™ Mono & Maxi Direct Stirrers
Ensure reliability, safety and performance with these mono and maxi direct stirrers, which are 100% maintenance- and wear-free.
Have Questions?
Catalog NumberModelSpeedVoltagePlug Type
50094713Maxi Direct80 to 2000 rpm100/240 VCord set w/various plugs
Catalog number 50094713
Price (SGD)
1,671.00
EA
Add to cart
Model:
Maxi Direct
Speed:
80 to 2000 rpm
Voltage:
100/240 V
Plug Type:
Cord set w/various plugs
Price (SGD)
1,671.00
EA
Add to cart
Ensure reliability, safety and performance with Thermo Scientific™ Cimarec™ Mono & Maxi Direct Stirrers which are 100% maintenance- and wear-free. With powerful magnets and no moving parts, these inductive-drive stirrers are the perfect choice for your general lab needs
Features:
  • Large, flat, easy-to-clean work surfaces
  • Compact, space-saving design
  • Gradual start acceleration is gentle and ensures optimum magnetic coupling
  • Adjustable power settings are separate from the speed control, decreasing power consumption and lowering heat output
  • Operating conditions are -10°C to +40°C at 95%RH
Mono Direct Stirrer:
  • Easy-to-use, rotary knob design
  • Rugged, powder-coated, stainless-steel housing
  • Stir speeds up to 1200rpm
  • Stir volumes up to 3L
  • IP32 protection rated
Maxi Direct Stirrers:
  • Precise microprocessor control with bright digital display
  • Easy-to-clean, chemical-resistant, sealed stainless-steel housing
  • Easily recall last used settings
  • Smooth and even stirring as low as 80rpm
  • Stir speeds up to 2000rpm
  • Stir volumes up to 5L
  • IP62 protection rated; easily cleaned with running water
Includes:
100-240V units contain a cord set with various plugs.
Warranty: 5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed80 to 2000 rpm
Stirring Positions1
Top Plate MaterialStainless Steel
Capacity5 L
DisplayDigital
ShapeSquare
Voltage100/240 V
Wattage5/10/15/21 w
Dimensions (L x W x H)8.5 x 7.1 x 1.4 in. (215 x 180 x 35 mm)
Height (English)1.4 in.
Height (Metric)35 mm
Length (English)8.5 in.
Length (Metric)215 mm
Width (English)7.1 in.
Width (Metric)180 mm
Plug TypeCord set w/various plugs
Certifications/ComplianceCE
ModelMaxi Direct
Protection ClassIP62, IP32
Temperature Range (Metric)-10°C to +40°C (Operating)
TypeStirrer
Weight (English)3.1 lb.
Weight (Metric)1.4 kg
Unit SizeEA