Cimarec™ i 20C Controller, 100-240V
Cimarec™ i 20C Controller, 100-240V
Actual product may vary
Cimarec™ i 20C Controller, 100-240V
Simplify operations with this user-friendly, menu-driven mircoprocessor controller included with the Micro, Mini, Compact, Maxi and Telesystems Stirrers.
Have Questions?
Catalog NumberLength (Metric)Type
5009077316.5 cmStirrer Controller
Catalog number 50090773
Price (SGD)
2,041.00
EA
Add to cart
Length (Metric):
16.5 cm
Type:
Stirrer Controller
Price (SGD)
2,041.00
EA
Add to cart
Simplify operations with the Thermo Scientific™ Cimarec™ i 20C Controller, a user-friendly, menu-driven microprocessor controller that is included with the Micro, Mini, Compact, Maxi and Telesystems Stirrers.
Features:
  • Single-handed operation with straightforward turn and press adjustment wheel
  • Three individual program, store and recall keys for rapid start-up
  • Last used settings stored in memory
  • Rocking/shake mode reverses stir bar direction for a gentle back and forth stirring motion
  • Four power settings to decrease power consumption and lower heat output
Ordering Information:
Order as replacement controller only.
Specifications
Height (English)3.7 in.
Height (Metric)9.5 cm
Length (English)6.4 in.
Length (Metric)16.5 cm
Pause Times5 sec. to 60 min.
Rated Power20 w
Speed130 to 1400 rpm
Speed Control Accuracy±1
Starting Times5 sec. to 60 min.
Temperature (Metric)0°C to 40°C @ 80% relative humidity
TypeStirrer Controller
Width (English)6.1 in.
Width (Metric)15.5 cm
Accuracy±1
Voltage100/240 V
Stirring Range130 RPM to 1400 RPM
Stirring Power Relative to Rated Power25/50/75/100 (4 levels)
Unit SizeEA