Cimarec™ i Mini Stirrers
Cimarec™ i Mini Stirrers
Actual product may vary
Cimarec™ i Mini Stirrers
Work in the smallest of places with these mini stirrers, which come with or without your choice of controller.
Have Questions?
Catalog NumberModeldimVoltagePlug Type
50088140Mini w/o controllerN.A.
Catalog number 50088140
Price (SGD)
684.00
EA
Add to cart
Model:
Mini w/o controller
Plug Type:
N.A.
Price (SGD)
684.00
EA
Add to cart
Work in the smallest of places with Thermo Scientific™ Cimarec™ i Mini Stirrers. The i Mini Series stirrers come with or without your choice of controller for use with the distribution system.
Features:
  • No larger than a thumbnail, perfect for stirring cuvettes
  • Stir speeds up to 1400rpm
  • Stir volumes up to 5mL
  • Operating conditions are -10°C to +56°C at 95%RH
  • Protection class IP 68 (stirrer only)
  • Link and Sync up to 8 mini stirrers using one controller with optional distributors and 50088140 stirrers
Ordering Information:
Choose from two different control units: the standard Telemodul controller with its space-saving design or the Telemodul 20 C with microprocessor controls, adjustable power setting and programmability.
Includes:
100-240V units contain a cord set with various plugs.
Warranty: 5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed1400 rpm
Capacity5 mL
ShapeSquare
Plug TypeN.A.
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
DescriptionMini w/o controller
ModelMini w/o controller
Temperature Range (Metric)-10°C to +56°C (Operating)
TypeStirrer
Unit SizeEA