Herasafe™ KS Options and Accessories
Herasafe™ KS Options and Accessories
Actual product may vary
Herasafe™ KS Options and Accessories
Worksurfaces, exhaust transitions and baskets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
50076162Vibration damped Worksurface 600 mm Width, for Weighing and Microscopy Equipment (316SS)
50073667Flat Worksurface Module 600 mm Wide (316SS)
50072685Flat Worksurface Module 300 mm Wide (316SS)
Catalog number 50076162
Price (SGD)
4,845.00
EA
Add to cart
Description:
Vibration damped Worksurface 600 mm Width, for Weighing and Microscopy Equipment (316SS)
Price (SGD)
4,845.00
EA
Add to cart
Worksurfaces, exhaust transitions and baskets
Specifications
DescriptionVibration damped Worksurface 600 mm Width, for Weighing and Microscopy Equipment (316SS)
Product TypeVibration Damped Worksurface
Width (Metric) Exterior600 mm
For Use With (Equipment)Biosafety Cabinet
Unit SizeEA