Heracell™ Support Frame with Casters
Heracell™ Support Frame with Casters
Actual product may vary
Heracell™ Support Frame with Casters
Heracell™ Support Frame with Casters
Heracell™ Support Frame with Casters
Heracell™ Support Frame with Casters
Heracell™ Support Frame with Casters
For Use With: Heracell 150i CO2 Incubator
Have Questions?
Catalog NumberdimVoltage
50057161
Catalog number 50057161
Price (SGD)
1,238.00
EA
Add to cart
Price (SGD)
1,238.00
EA
Add to cart
For Use With: Heracell 150i CO2 Incubator
  • Provide protection against floor contamination
  • Choice of three heights: 185mm (7.3 in.), 200mm (7.9 in.), or 780mm (30.7 in.)
  • Support frames can also be caster-mounted for easy maneuverability
Specifications
For Use With (Equipment)Heracell 150i CO2 incubator
LightingLED
TypeSupport Frame
IncludesCaster
Height (English)7.3 in.
Height (Metric)18.5 cm
Unit SizeEA