Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Sterility |
---|---|---|
5000-1012 | 1.0 mL | Sterile |
FIS03-337-7C | 1.2 mL | Sterile |
FIS03-337-7Y | 1.5 mL | Sterile |
5000-0020 | 2.0 mL | Sterile |
FIS03-337-7D | 2.0 mL | Sterile |
FKS11750573 | 2.0 mL | Sterile |
5000-0050 also known as NLG5000-0050 | 5.0 mL | Sterile |
FIS03-337-7H | 5.0 mL | Sterile |
FKS11365644 | 5.0 mL | Sterile |
FIS03-337-7B | 15 mL | Sterile |
5011-0020 also known as RBIN5011-0020 | 2.0 mL | Non-sterile |
FIS03-337-7P | 2.0 mL | Non-sterile |
5012-0020 | 2.0 mL | Sterile |
Enhance aseptic techniques with externally threaded Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes. These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to vapor phase liquid nitrogen temperatures.