Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Actual product may vary
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts
Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
Have Questions?
Catalog NumberColorProduct Type
4906YellowScrew Cap Tray, Empty
Catalog number 4906
HSN 39269099
Price (SGD)
104.00
CS
Color:
Yellow
Product Type:
Screw Cap Tray, Empty
Price (SGD)
104.00
CS
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Highest Quality
  • All caps and inserts are supplied free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Superior Cap Design
  • Gasketed caps ensure critical sample protection over the storage lifetime
  • Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
  • Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
  • ScrewTop caps are compatible with storage down to -176°C
Cap Options Simplify Sample Identification
  • Cap trays feature standard microplate format compatible with automated equipment
  • Caps are available in seven color options to enable sample identification
  • Color inserts snap into cap for a secure fit; inserts are removable
  • Inserts are available in six colors
Specifications
Product TypeScrew Cap Tray, Empty
MaterialABS
ColorYellow
Packaging5 Empty Trays for 96 Caps
Product LineMatrix
SterilitySterile, Sterile
Pore Size0.2 μm
Fit for PurposeSterile and PCR Ready
Unit SizeCS