Laboratory Manifold MicroFunnel™ Adapter
Laboratory Manifold MicroFunnel™ Adapter
Actual product may vary
Laboratory Manifold MicroFunnel™ Adapter
Accessory adapter for MicroFunnel filter funnels.
Have Questions?
Catalog number 4890
Price (SGD)
261.00
CS
Add to cart
Price (SGD)
261.00
CS
Add to cart
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifications
SterilityNon-sterile
No. per Pack3/Pk.
TypeMicroFunnel Adapter
Unit SizeCS
Frequently asked questions (FAQs)
Which racks are suitable for storage of the tubes in which TaqMan assays are delivered?
The Racks for Matrix 2D Barcoded Storage Tubes (Cat. No. 4890) are suitable for storage of narrower tubes. For more products and catalog numbers, see the following link.
Find additional tips, troubleshooting help, and resources within our TaqMan® Primers and Probes Support Center
Can I store the Thermo Scientific 2D barcoded storage tubes in standard 10x10 cryoboxes?
Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.