1x1 image pixel for data collection
Immuno Tubes and Stoppers
Immuno Tubes and Stoppers
Actual product may vary
Immuno Tubes and Stoppers
Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)MaterialDiameter (Metric) OuterProduct Type
4703195 mLPolystyrene12 mmImmuno Star Tubes and Stopper
4442025 mLPolystyrene12 mmImmuno Tubes and Stopper
Catalog number 470319
Price (SGD)
600.00
CS
Add to cart
Volume (Metric):
5 mL
Material:
Polystyrene
Diameter (Metric) Outer:
12 mm
Product Type:
Immuno Star Tubes and Stopper
Price (SGD)
600.00
CS
Add to cart
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:
  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment
Choice of Surface:
  • Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
  • Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
  • Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • High–quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times
Recommended for:
  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
Specifications
MaterialPolystyrene
Product TypeImmuno Star Tubes and Stopper
SurfaceMaxiSorp
Length (Metric)75 mm
Diameter (Metric) Outer12 mm
Certifications/ComplianceMaxiSorp tubes are certified for IgG binding (tube to tube % CV less than 5%)
No. per Pack100
No. per Case3000
Volume (Metric)5 mL
For Use With (Equipment)Immuno Tube
Unit SizeCS
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link