1x1 image pixel for data collection
Finnpipette™ Novus GLP Kits
Finnpipette™ Novus GLP Kits
Actual product may vary
Finnpipette™ Novus GLP Kits
Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.
Have Questions?
Catalog NumberVolume (Metric)
47011201 to 1000 μL
Catalog number 4701120
Price (SGD)
2,229.00
EA
Add to cart
Volume (Metric):
1 to 1000 μL
Price (SGD)
2,229.00
EA
Add to cart
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
Novus GLP Kit 1 includes:
  • Finnpipette Novus 10-100μL
  • Finnpipette Novus 100-1000μL
  • Two Finnpipette Novus stands
  • One rack of Finntip Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Finnpipette Novus brochure
Novus GLP Kit 2 includes:
  • Finnpipette Novus 100-1000μL
  • Finnpipette Novus 0.5-5mL
  • Two Finnpipette Novus stands
  • One rack of Finntip 5ml and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Finnpipette Novus brochure
  • Pipetting Guide
Novus GLP Kit 3 includes:
  • Novus pipette, 1-10μL, micro
  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 10, Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure
Novus GLP Kit 4 includes:
  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Novus pipette, 1-10mL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 200, Flex 1000 and Finntip 10ml
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure
Specifications
ModelNovus
Number of Channels1
Product LineNovus
TypeGLP Kit
Volume (Metric)1 to 1000 μL
IncludesNovus 10micro, 100 and 1000 μL single channel pipettes, three Novus stands, three sample Finntip racks
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link