Finnpipette™ F3 Multichannel Channel Pipettes
Finnpipette™ F3 Multichannel Channel Pipettes
Actual product may vary
Finnpipette™ F3 Multichannel Channel Pipettes
Multichannel pipettes that provide comfort in everyday pipetting.
Have Questions?
Catalog NumberVolume (Metric)Number of Channels
466002030 to 300 μL8
Catalog number 4660020
Price (SGD)
1,214.00
EA
Add to cart
Volume (Metric):
30 to 300 μL
Number of Channels:
8
Price (SGD)
1,214.00
EA
Add to cart
Thermo Scientific™ Finnpipette™ F3 Multichannel Pipettes are comfortable to use, and provide ideal functionality in microplate applications. A wide, color-coded supportive finger rest and ergonomic handle design will enhance your pipetting experience. Finnpipette F3 pipettes are available in single and multichannel models.
Ergonomic Comfort
  • Wide color-coded supportive finger rest for identification support and ergonomics
  • Designed handle softly contours the shape of your hand
  • Large Ergovisio Display for volume recognition
Ideal Function
  • Durable handle material
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Advanced volume gearing mechanism for enhanced precision and accuracy
  • Super blow-out for efficient delivery of micro-size drops in 50μL models and below
Specifications
Accuracy±5.00 to 1.0%
Compatible TipsFinntip 300, Filter 300, Flex 300, Flex Filter 300
DescriptionFinnpipette F3, 8-ch,  30-300μL
AutoclavableAutoclavable
ColorOrange
ModelF3
Number of Channels8
Product LineFinnpipette
TypePipette
Volume (Metric)30 to 300 μL
Increments1μL
Precision2.0 to 0.3%
Color CodeOrange
Unit SizeEA