Finnpipette™ F1 Fixed Volume Single-Channel Pipettes
Finnpipette™ F1 Fixed Volume Single-Channel Pipettes
Actual product may vary
Finnpipette™ F1 Fixed Volume Single-Channel Pipettes
Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.
Have Questions?
Catalog NumberVolume (Metric)AccuracyPrecision
465102010 μL+/-0.9%0.8%
Catalog number 4651020
Price (SGD)
-
Volume (Metric):
10 μL
Accuracy:
+/-0.9%
Precision:
0.8%
Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50μL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.
Features:
  • Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
  • Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
  • Light, smooth pipetting action: Enables better results over longer pipetting periods
  • Super blow-out piston with volumes of 50μL and below: Ensures delivery of micro-size drops
  • Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection
Warranty: 5 years with web registration
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy+/-0.9%
DescriptionF1 Fixed Volume Single Channel
For Use WithFinnTip™ Flex 200, 250 Univ., 200 Ext
ModelF1
Number of Channels1
TypeFixed Volume Pipette
Volume (Metric)10 μL
Precision0.8%
Unit SizeEach