Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Actual product may vary
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Finnpipette™ Digital Variable Volume Single Channel Pipettes
Comfortably perform daily pipetting tasks with these reliable and durable manual variable volume pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color Code
450-00200.5 to 10 μLGray
45001005 to 50 μLOrange
Catalog number 450-0020
Price (SGD)
329.00
CS
Add to cart
Volume (Metric):
0.5 to 10 μL
Color Code:
Gray
Price (SGD)
329.00
CS
Add to cart
Thermo Scientific™ Finnpipette™ Digital Variable Volume Single Channel Pipettes feature a classic style for timeless dependability and durability. The Finnpipette Digital pipette is fully autoclavable, and combines robust durability with consistent performance. Finnpipette Digital pipettes are available in single and multichannel models.
Consistent Performance
  • Fully autoclavable
  • No metal parts to rust or corrode
  • Delivers micro-size drops with the patented super blow-out piston in 50μL models and below
Comfort and Ease of Use
  • Easily set volume by t urning the large operating button
  • Quickly read volume selection in large digital window display
  • Gearing mechanism releases tips with a gentle push
Specifications
Accuracy±3.5 to 1%
Compatible TipsFinntip Flex 200, 200 Ext, 250 Univ, QSP tip 200, 300
AutoclavableAutoclavable
ColorGray
For Use WithFinnTip™: Flex 200, 200 Extended, 250 Universal, QSP™ Tip: 200, 300
ModelFinnpipette™ Digital
Number of Channels1
Product LineFinnpipette
DisposableSingle-use
SterilitySterile, Sterile
TypePipette
Volume (Metric)0.5 to 10 μL
Material (Membrane)Cellulose Nitrate
Pore Size0.2 μm
Increments0.1 μL
Precision3 to 0.8%
Grid ColorWhite/None
No. per Case12
No. per Pack12
Color CodeGray
TypeFilter Unit
Volume (Metric) Receiver500 mL
Volume (Metric) Upper500 mL
Diameter (Metric) Membrane75 mm
Fit for PurposeSterile
Unit SizeCS
Frequently asked questions (FAQs)
What is the maximum volume that I can run through the Nalgene Rapid-Flow Sterile Disposable Filter Units with PES, CN, SFCA or Nylon Membranes?
The filters included with Nalgene Rapid-Flow Sterile Single Use Vacuum Filter Units are not specifically rated for a maximum volume that may be filtered. In many cases it depends on the specific application of the filter unit and would require testing beyond the volume of the receiver. Signs of the filter clogging, such as a decrease in filtration rate, would likely indicate the need for replacement.
I am using a Nalgene Rapid-Flow Sterile Disposbable Filter Unit and my solution is taking a long time to filter. Do you have any recommendations to improve filtering?
We recommend using glass fiber prefilters for filtration of solutions with high viscosity or especially high particulate load. The glass fiber prefilters are available as an optional accessory and bundled with select Thermo Scientific Nalgene Rapid-Flow filter units. The prefilters enable a higher volume to be filtered before significant flow decay by trapping and preventing the larger particulate matter from entering the final filter membrane. Simply place one prefilter on top of the membrane bonded inside the unit, wet slightly to hold in place, and proceed with filtration as usual.