KLEENEX™ 27 Toilet Tissue Bulk Pack
KLEENEX™ 27 Toilet Tissue Bulk Pack
Actual product may vary
KLEENEX™ 27 Toilet Tissue Bulk Pack
KLEENEX™ 27 Toilet Tissue Bulk Pack
KLEENEX™ 27 Toilet Tissue Bulk Pack
Dispensed one at a time so they are touched only by the user. Kimberly-Clark™ KLEENEX™ 27 Toilet Tissue Bulk Pack tissues dissolve in cold water to reduce blocked drains.
Have Questions?
Catalog NumberDimensions (L x W)
4477
also known as 4477PIN
18.6 x 12.5 cm
Catalog number 4477
also known as 4477PIN
Price (SGD)
451.00
CS
Add to cart
Dimensions (L x W):
18.6 x 12.5 cm
Price (SGD)
451.00
CS
Add to cart
  • Supplied in packs of 27 x 260 white, 2-ply folded toilet tissue sheets
  • Embossed with the Kleenex™ brand
  • Dissolve in water, preventing blockages and reducing maintenance
  • Manufactured from 100% recycled, FSC-certified paper paper
  • Dispensed one at a time so they are touched only by the user
  • Help reduce costs and waste with up to 40% less used compared to rolls
  • Bulk packaging helps to reduce contamination and the spread of germs
  • ISO 14001, ISO 9001, EU Ecolabel and FSC certified
  • Ideal for office and hotel washrooms that have a high-quality image
  • For use with the wall-mounted AQUARIUS™ Toilet Tissue Dispenser
Specifications
Product TypeToilet Tissue
Length (English)11.54 in.
Length (Metric)18.6 cm
Width (Metric)12.5 cm
MaterialPaper
Certifications/ComplianceISO 14001, ISO 9001, EU Ecolabel, FSC certified
ColorWhite
Dimensions (L x W)18.6 x 12.5 cm
FormatFolded
For Use With (Application)Ideal for office and hotel washrooms that have a high-quality image
Packaging Quantity27 x 260 Sheets
Ply2-ply
Unit SizeCS