Acro™ 37 TF Vent Device with PTFE Membrane
Acro™ 37 TF Vent Device with PTFE Membrane
Actual product may vary
Acro™ 37 TF Vent Device with PTFE Membrane
Disposable, multipurpose filters for small volume venting, solvent, and gas filtration.
Have Questions?
Catalog NumberQuantity
4465200/Pk.
Catalog number 4465
Price (SGD)
136.00
CS
Add to cart
Quantity:
200/Pk.
Price (SGD)
136.00
CS
Add to cart
  • PTFE membrane and polypropylene housing have broad chemical compatibility.
  • This product is not approved for use in medical, clinical, surgical or other patient applications.
Specifications
Material (Membrane)PTFE
Pore Size0.2 μm
SterilityNon-sterile
Filtration Area7.5 cm2
Quantity200/Pk.
Diameter (Metric)37 mm
Unit SizeCS
Frequently asked questions (FAQs)
What are the main advantages of the Thermo Scientific 2D barcoded storage system?
Here are some advantages of the 2D barcoded storage system:
- Less time needed for labeling, sample recording, and retrieval
- Maximize freezer utilization while keeping storage space organised
- Ensures a safe, permanent ID of a sample
- Enables multivial capping and multichannel filling
- Individualized samples, eliminating multiple freeze/thaw cycles
Which Thermo Scientific 2D barcoded storage tubes should I use for my application?
Please view the following information regarding the usage of Thermo Scientific 2D barcoded storage tubes:
Tube Type; Typical Storage Temperature; Application(s); Special Feature(s)
Sepraseal; -20 degrees C; Compound storage; Can easily be removed for sample access
Presplit Sepraseal; -20 degrees C; Compound storage; Up to 200 piercings
Duraseal; -20 degrees; Compound storage; Safe seal for multiple piercings
Matrix ScrewTop; -80 degrees C; Blood components, DNA, Bacteria; With the correct torque, there is very little evaporation at -80 C, disposable pipette tips can be used
Nunc Screwcap; -176 degrees C; Mammalian cells; Minimal height, tubes can be removed with cryo gloves