Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Actual product may vary
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentWell ShapeNo. per PackProduct Type
446469
also known as 44646 9
MaxiSorpRound, C Bottom10Strip Plate
FIS22-047027PolySorpRound, C Bottom5Strip Plate
446639MaxiSorpRound10Microplate Strip
Catalog number 446469
also known as 44646 9
Price (SGD)
977.00
CS
Add to cart
Surface Treatment:
MaxiSorp
Well Shape:
Round, C Bottom
No. per Pack:
10
Product Type:
Strip Plate
Price (SGD)
977.00
CS
Add to cart
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
  • Free-standing modules are easy to handle
Specifications
BreakapartBreakable Strips (LockWell)
Binding PropertyHydrophilic
CertifiedYes
FormatStrip Plate
LidWithout Lid
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
Array8 x 12
Packaging10 Pk.
ColorClear
Product TypeStrip Plate
MaterialPolystyrene
Volume (Metric) Well350 μL
No. of Wells96
Volume (Metric) Working250 μL
SterilityNon-sterile
Surface TreatmentMaxiSorp
Well ShapeRound, C Bottom
For Use With (Application)For antibody sandwich assays
No. per Case60
No. per Pack10
Plate BlockingNone
Quantity60/Cs.
Target MoleculeAntibodies (Glycoproteins)
Well DesignC8
Unit SizeCS
Frequently asked questions (FAQs)
What length of peptide is ideal for binding to the Thermo Scientific MaxiSorp surface? What are the detection limitations?
We have tested and found that a 3 amino acid peptide (Pro, Leu, Gly) cannot be detected when passively adsorbed on the MaxiSorp surface. However, this peptide can be detected when covalently immobilized using CovaLink NH Modules and CovaLink NH2 Modules and Plates. Using covalent immobilization of small peptide residues, you can typically obtain a better orientation of the molecule and reduced problems with antibody recognition of the peptide due to masking of the epitope. We have discovered that a 7 amino acid peptide from the MHC Class II antigen can be detected when adsorbed on the MaxiSorp surface. We state that the detection limitation using the MaxiSorp surface is between 3 and 7 amino acid residues.
One additional note is that detection is contingent upon the orientation of the peptide when immobilized. If the active site is inactivated or hidden at the site facing the solid phase, no detection signal is observed.
Can I bind either single- or double-stranded DNA to the Thermo Scientific MaxiSorp surface?
Single-stranded DNA can be adsorbed to our MaxiSorp surface using approximately 10 µg ssDNA per mL PBS, pH 8.2, although the stability is uncertain. Based on our experience, ssDNA immobilized on the MaxiSorp surface is so loosely bound that it can be removed by stringent washing.
Double-stranded DNA will not bind to the MaxiSorp surface. DNA, however, can be covalently bound to Nunc NucleoLink Strips.
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?
Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.
For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?
The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.
What is the difference between certified and non-certified MaxiSorp plates and modules?
Both of these surfaces are identical. The only difference between them is that for the certified plates, a representative sample from each manufacturing lot undergoes a Binding Capacity test. This test is an ELISA-like assay used in our quality control laboratories to ensure binding capabilities.