Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Tin(II) chloride dihydrate, 98+%, ACS reagent
Acros Organics
Tin(II) chloride dihydrate, 98+%, ACS reagent
CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Catalog NumberQuantityPackaging
424481000100 gPlastic Bottle
Catalog number 424481000
HSN 28273910
Price (SGD)
-
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder, crystals and chunks
Titration Iodimetric98.0 to 103.0 %
Sulfate (SO4)Passes test (limit about 0.003%)
Calcium (Ca)=<50 ppm