DeckWorks™ Tip Stations, Nonsterile
DeckWorks™ Tip Stations, Nonsterile
Actual product may vary
DeckWorks™ Tip Stations, Nonsterile
Designed for single and multichannel pipetting stations
Have Questions?
Catalog NumberVolume (Metric)
4141
also known as USFSS4141
0.1 to 10 μL
Catalog number 4141
also known as USFSS4141
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
0.1 to 10 μL
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip ­stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and ­post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
Volume (Metric)0.1 to 10 μL
SterilityNon-sterile
ColorNatural
Tip StyleRound
PackagingMulti-pack
For Use With (Equipment)Universal Fit, Multichannel Pipetting Stations
GraduatedYes
Product LineDeckWorks