8-Channel Screw Cap Decapper
8-Channel Screw Cap Decapper
Actual product may vary
8-Channel Screw Cap Decapper
Process a full rack of screw-capped lab tubes in less than one minute with this ergonomic, manual capper/decapper
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
4105NUNCapper/DecapperNunc-style Tubes
4105MATCapper/DecapperMatrix-style Tubes
Catalog number 4105NUN
Price (SGD)
4,919.00
EA
Add to cart
Type:
Capper/Decapper
For Use With (Equipment):
Nunc-style Tubes
Price (SGD)
4,919.00
EA
Add to cart
Process a full rack of Thermo Scientific™ Matrix™ or Thermo Scientific™ Nunc™ tubes in less than one minute with this ergonomic, manual capper/decapper. The Thermo Scientific™ 8-Channel Screw Cap Capper/Decapper is designed to cap tubes to the optimal torque to maintain sample integrity and prevent sample loss during storage. The 8-channel Capper/Decapper is a practical addition to benchtop processing of screw-top storage tubes.
Warranty: 1 year
Specifications
Compatible Storage TubesNunc-Style Tubes
Decapping Format8-Channel
Description8-Channel Handheld Screw Cap Capper/Decapper with Stand for Nunc-style Tubes
Dimensions (L x W x H)9.87 x 3.76 x 2 in. (25.1 x 9.5 x 5 cm)
Electrical Requirements110 to 240V 50/60Hz
Height (English)2 in.
Height (Metric)5 cm
Length (English)9.87 in.
Length (Metric)25.1 cm
Net Weight (English)0.97 lb.
Net Weight (Metric)0.44 kg
No. per Case1
No. per Pack1
OperationManual via Toggle Switch
Recharge Time90 min.
Voltage110/240 V
Width (English)3.76 in.
Width (Metric)9.5 cm
Battery Run TimeMinimum 20 Racks, Cap and Decap
TypeCapper/Decapper
For Use With (Equipment)Nunc-style Tubes
Frequency50/60 Hz
Speed4 sec. to Cap/Decap 8 Tubes
Unit SizeEA