Check Order Status
Cryopreservation Vials, Canes and Canisters
Actual product may vary
Cryopreservation Vials, Canes and Canisters
Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.
Have Questions?
Catalog Number
Type
For Use With (Equipment)
4000211
also known as 4000OPQ211
Cryo Cane
6 Ampules
Catalog number
4000211
also known as 4000OPQ211
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Type:
Cryo Cane
For Use With (Equipment):
6 Ampules
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
Warranty varies by country, please contact us for details
Specifications
Type
Cryo Cane
Capacity (English)
4.23 cu. ft.
Capacity (Metric)
1.2 mL
Color
Silver
For Use With (Equipment)
6 Ampules
Holds
Ampules (6)