CryoPlus™ Vapor Phase Platform Risers and Frames
CryoPlus™ Vapor Phase Platform Risers and Frames
Actual product may vary
CryoPlus™ Vapor Phase Platform Risers and Frames
Incorporate Thermo Scientific™ CryoPlus™ Vapor Phase Platform Risers and Frames with CryoPlus™ LN2 cryopreservation storage systems for a complete storage solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Height (English) Exterior
4000062CryoPlus 2, Rack 40000054 in.
4000056CryoPlus 1, Rack 40000054 in.
Catalog number 4000062
Price (SGD)
449.00
EA
Add to cart
For Use With (Equipment):
CryoPlus 2, Rack 4000005
Height (English) Exterior:
4 in.
Price (SGD)
449.00
EA
Add to cart
Incorporate Thermo Scientific™ CryoPlus™ Vapor Phase Platform Risers and Frames with CryoPlus™ LN2 cryopreservation storage systems for a complete storage solution.
Vertical
  • 5.0 inch riser for rack model no. 4000011
Square
  • 3.5 inch riser for rack model no. 4000001
  • 3.5 inch riser for rack model no. 4000002
  • 4.0 inch riser for rack model no. 4000005
  • 5.0 inch riser for rack model no 4000003
  • 5.0 inch riser for rack model no. 4000004
SUC-1 (Canister Cane Storage)
  • 3.5 inch riser for rack model no. 4000379
Arrowhead
  • 4.0 inch riser for rack model no. 4000097
  • 4.0 inch riser for rack model no. 4000100
  • 4.0 inch riser for rack model no. 4000101
  • 4.0 inch riser for rack model no. 4000108
  • 4.0 inch riser for rack model no. 4000109
  • 4.0 inch riser for rack model no. 4000110
  • 4.0 inch riser for rack model no. 4000111
Frames
  • Bag capacity 50mL, Fenwal 4R9951, 6 places for cassettes
  • Bag capacity, 250mL, Fenwal 4R5461, 4 places for cassettes
  • Bag capacity, 500mL, Fenwal 4R5462, 4 places for cassettes
  • Warranty varies by country, please contact us for details
Specifications
TypeRiser
Capacity (Metric)500 mL
For Use With (Equipment)CryoPlus 2, Rack 4000005
Height (English) Exterior4 in.
HoldsRacks
Unit SizeEA