EUROSTAR 200 Digital Overhead Stirrer
EUROSTAR 200 Digital Overhead Stirrer
Actual product may vary
EUROSTAR 200 Digital Overhead Stirrer
Extremely powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100 L (H2O).
Have Questions?
Catalog NumberSpeed
39900016 to 2000 rpm
Catalog number 3990001
Price (SGD)
2,188.00
EA
Add to cart
Speed:
6 to 2000 rpm
Price (SGD)
2,188.00
EA
Add to cart
  • Automatically adjusts the speed through microprocessor controlled technology within the speed range
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
Specifications
Speed6 to 2000 rpm
Capacity100 L
DisplayLED
Voltage100/115 V
Wattage130 W
Dimensions (L x W x H)209 x 91 x 274 mm (8.2 x 3.6 x 10.8 in.)
Height (English)10.8 in.
Height (Metric)274 mm
Length (English)8.2 in.
Length (Metric)209 mm
Width (English)3.6 in.
Width (Metric)91 mm
Frequency50/60 Hz
ModelEUROSTAR 200
Temperature Range (Metric)5°C to 40°C
Torque200 Ncm
TypeOverhead Stirrer
Viscosity100,000 mPas
Unit SizeEA