Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks
Use the automation friendly tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Have Questions?
Change view
Catalog Number
Barcode
Packaging
374502
2D on Tube Bottom
Bulk
FKS13416113
2D on Tube Bottom
Bulk
2 Options
Catalog number 374502
HSN 39269099
Price (SGD)
1,084.00
CS
Barcode:
2D on Tube Bottom
Packaging:
Bulk
Price (SGD)
1,084.00
CS
Use the automation friendly Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.
Scalable
Tubes and rack are ideal for both lower-throughput and high-throughput use
Feature graduations and white patch for manual marking
Can be used in traditional cryostorage boxes made for 10x10 arrays
External threads limit contamination
Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
Single-piece construction with molded-in gasket
Cannot be over-tightened manually or with automated capping tools
Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
Scan the tube to access sample information and identify the contents
374502NOV is featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube
Stringent Quality Control
Every 2D barcoded storage tube is scanned to ensure readability
Barcodes are checked against database of previously-assigned codes to prevent duplicates
Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Specifications
ColorNatural
Product TypeExternally-threaded Universal Tube
SterilitySterile, Sterile
DescriptionNunc Universal tubes with 2D code
Closure ColorNatural
Capacity (Metric)1.8 mL
Bottom ShapeFlat
MaterialPlastic
Thread StyleExternal
For Use With (Equipment)Cryobank
IncludesWhite patch and graduations
No. per Case480/Cs.
PackagingBulk
Barcode2D on Tube Bottom
Unit SizeCS
Frequently asked questions (FAQs)
What is the Thermo Fisher alternative to Corning® 2 mL Internal Threaded Polypropylene Cryogenic Vial with 1D and 2D Bar codes, Product Number 8670?
The following alternatives for the Corning Cryogenic Vial, Product Number 8670:
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374510, 374510-BR)
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374512). Please note that this product is FDA registered and CE-marked.
- Nunc 1.8mL Externally-Threaded Universal Tubes and Racks (Cat. No. 374502, 374502NOV
- Matrix ScrewTop Tri-coded Tubes (Cat. No. 3740-WP1D)