Connexin 47 Monoclonal Antibody (4A11A2), Invitrogen™
Connexin 47 Monoclonal Antibody (4A11A2), Invitrogen™
Actual product may vary
Connexin 47 Monoclonal Antibody (4A11A2), Invitrogen™
Connexin 47 Monoclonal Antibody (4A11A2), Invitrogen™
Connexin 47 Monoclonal Antibody (4A11A2), Invitrogen™
Mouse Monoclonal Antibody
Have Questions?
Catalog NumberQuantity
374500
also known as S374-500
100 μg
Catalog number 374500
also known as S374-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 μg
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
AntigenConnexin 47
ApplicationsImmunohistochemistry, Immunoprecipitation, Western Blot
ClassificationMonoclonal
Clone4A11A2
Concentration0.5 mg/mL
ConjugateUnconjugated
FormulationPBS with 0.1% sodium azide; pH 7.4
GeneGJC2
Gene Accession No.Q8BQU6
Gene AliasB230382L12Rik; connexin 47 type A; connexin 47 type C; connexin 47 type D; connexin-46.6; Connexin-47; CX46.6; Cx47; CXG2; gap junction alpha-12 protein; Gap junction gamma-2 protein; gap junction membrane channel protein alpha 12; gap junction protein gamma 2; gap junction protein, alpha 12; gap junction protein, chi 2; gap junction protein, gamma 2; gap junction protein, gamma 2, 47kDa; GJA12; Gjc2; HLD2; LMPH1C; PMLDAR; SPG44
Gene SymbolsGJC2
Host SpeciesMouse
ImmunogenSynthetic peptide derived from the C-terminal region of the mouse Connexin 47 protein
Purification MethodProtein A
Quantity100 μg
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)118454
Target SpeciesMouse
Product TypeAntibody
FormLiquid
IsotypeIgG1 κ
Unit SizeCS
Contents & Storage
-20°C