Forma&trade; Steri-Cycle&trade; CO<sub>2</sub> Incubator, 184 L, Polished Stainless Steel
Actual product may vary
Forma&trade; Steri-Cycle&trade; CO<sub>2</sub> Incubator, 184 L, Polished Stainless Steel
Forma&trade; Steri-Cycle&trade; CO<sub>2</sub> Incubator, 184 L, Polished Stainless Steel
Forma™ Steri-Cycle™ CO2 Incubator, 184 L, Polished Stainless Steel
Experience continuous protection against unwanted airborne contaminants, and an on-demand, high-temperature sterilization cycle that simplifies routine cleaning.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltageCO2 Sensor Technology
371
also known as MIR371DAS
230 VTC Sensor
381230 VIR Sensor
Catalog number 371
also known as MIR371DAS
HSN 84198990
Price (SGD)
8,324.00
Special offer
Ends: 30-Jun-2026
9,792.00
Save 1,468.00 (15%)
Each
Voltage:
230 V
CO2 Sensor Technology:
TC Sensor
Price (SGD)
8,324.00
Special offer
Ends: 30-Jun-2026
9,792.00
Save 1,468.00 (15%)
Each
Experience complete contamination control and dependable long-term performance. Thermo Scientific™ Steri-Cycle CO2 Incubators offer precise direct-heat temperature control with choice of TC (thermal conductivity) or IR (infrared) sensor and convenient high-temperature decontamination cycle. Our reliable direct-heat Steri-Cycle CO2 incubator features a unique in-chamber HEPA air filtration system—which provides continuous protection against unwanted airborne contaminants—and an on-demand, high-temperature sterilization cycle that simplifies routine cleaning.
Performance
  • Approximate cycle length: 14 hr.
  • Optimized airflow system will not interfere with samples or incubator function
  • Optional built-in VOC filtration system removes volatile organic vapors
  • In-chamber HEPA air filtration continuously removes particulates for Class 100 quality conditions
  • On-demand 140°C sterilization cycle eliminates contamination from internal surfaces
  • Audible alarm activates if outer door is opened during the cycle or if temperature reaches 60°C (140°F) or greater
  • Heat-resistant TC sensor can be left in place during the cycle
  • HEPA filter and IR sensor are easily removed
  • Access code prevents accidental initiation of the cycle
Microprocessor Control
  • Alphanumeric messaging center clearly displays temperature, CO2 and (optional) RH parameters
  • Audio/visual alarms with silence button
  • Setpoints for temperature, overtemperature, CO2
  • Configurable audible ON/OFF, access code, HEPA filter change reminder, remote alarm contacts, tracking low temp and high and low CO2 alarms
Construction
  • Polished stainless-steel interior with coved corners is easy to clean
  • Sturdy stainless-steel shelves and supports can be removed without tools
  • Microbiological filters on gas inlet and sample ports are easily accessible
  • Removable inner door gasket adjusts continually to ensure tight seal
Ordering Information:
Options include: choice of CO2 sensors, reversible door swings, relative humidity monitoring, antimicrobial copper interior, automatic gas tank switchers, datalogging capability, IQ/OQ documentation kits.
Specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsOptional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Humidity SourceWater Pan
Dimensions (D x W x H) Interior20 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26.3 x 39.5 in. (63.5 x 66.8 x 100.3 cm)
Weight (English)260 lb.
Electrical Requirements230 V, 50/60 Hz