GC Syringes for TriPlus™ RSH Autosampler
Actual product may vary
GC Syringes for TriPlus™ RSH Autosampler
GC Syringes for TriPlus™ RSH Autosampler
GC Syringes for TriPlus™ RSH Autosampler
GC Syringes for TriPlus™ RSH Autosampler
Easily accomplish accurate, precise, and reproducible injections with Thermo Scientific™ GC Syringes for Thermo Scientific™ TriPlus™ RSH Autosampler.
Have Questions?
Catalog NumberVolume (Metric)
365D029110 μL
Catalog number 365D0291
HSN 90183100
Price (SGD)
-
Volume (Metric):
10 μL
Achieve accurate, precise, and reproducible injections easily using Thermo Scientific™ GC Syringes for Thermo Scientific™ TriPlus™ RSH Autosampler. The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes.
  • Syringe volumes from 0.5μL to 5mL
  • Gas-tight options available
Specifications
Description10 μL FN, 26s G, 57 mm
Needle Gauge26s G
Length (Metric)57 mm
Product TypeGC Autosampler Syringe
QuantityEa.
Volume (Metric)10 μL
Length (Metric) Needle57 mm
Needle Point StyleCone
For Use With (Equipment)TriPlus RSH Autosampler
PTFE Tipped PlungerNo
Unit SizeEach