Vacuum Ovens
Vacuum Ovens
Actual product may vary
Vacuum Ovens
Vacuum Ovens
Vacuum Ovens
Achieve maximum temperatures of 220°C (428°F) with this flexible oven, while benefiting from two control configurations and display options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionCapacity (Metric)
3618-1CE2.3 cu. ft. (65.1L); 240V 1600w 6.7A; Dial display65.1 L
3606-1CE0.44 cu. ft. (1.25L); 240V 600w 2.5A; Dial display12.5 L
3608-1CE0.7 cu. ft. (19.8L); 240V 600w 2.5A; Dial display19.8 L
3608-6CE19.8L (0.7 cu. ft.); 240V; 600w; 2.5A; LED display19.8 L
3618-6CE2.3 cu. ft.(65.1L); 240V 1600w 6.7A; LED display65.1 L
Catalog number 3618-1CE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
2.3 cu. ft. (65.1L); 240V 1600w 6.7A; Dial display
Capacity (Metric):
65.1 L
Achieve maximum temperatures of 220°C (428°F) with Thermo Scientific™ Vacuum Ovens, while benefiting from two control configurations and display options. These ovens feature independent vacuum and purge needle valves. Radiant warm-wall heating system optimizes uniformity and conserves chamber space for drying, curing, vacuum embedding and plating applications.
Features:
  • 3 convenient chamber sizes: 12.5L (0.44 cu.ft.), 19.8L (0.7 cu.ft.), 65.1L (2.3 cu.ft.)
  • Temperature Range: Ambient +10° to 220°C
  • Hydraulic thermostat temperature control with dial thermometer (5°C increments); medium and large units available with LED display (1°C increments)
  • Polycarbonate safety shield protects door glass
  • Silicone door gasket and positive latch door maintain seal at all vacuum levels – use of vacuum grease is recommended
  • Vacuum and air lines are corrosion-resistant, stainless-steel tubing for optimum chamber cleanliness and long-term performance
  • Two removable stacking aluminum shelves
  • Powder-coated, heavy-gauge steel exterior for durability
  • Radiant warm-wall heating system helps optimize uniformity and conserve chamber space; 3 in.(7.6cm) glass wool insulation prevents heat loss
Small 3606 Models
Top-mounted, independent evacuation and venting vacuum fittings
Medium and large 3608 and 3618 Models
Front-mounted, three-way valve for evacuation, venting and purging of inert gases (e.g. Nitrogen or Argon) with vacuum fittings on the front of the unit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)2.3 cu.ft.
Chamber MaterialStainless Steel
Description2.3 cu. ft. (65.1L); 240V 1600w 6.7A; Dial display
Capacity (Metric)65.1 L
Temperature (Metric)Ambient +10° to 220°C
TypeOTP
DisplayBimetallic Thermometer with 5°C increments
MaterialPowder-coated Heavy-gauge Steel
No. of Shelves2
Shelf MaterialAluminum
Temperature Resolution±1.0°C
Temperature Uniformity±5.0°C at 100°C 25 ft.Hg
Vacuum Range0 to 30 in.Hg
Dimensions (D x W x H) Interior20 x 14 x 14 in. (51 x 36 x 36 cm)
Dimensions (L x W x H)25 x 25 x 22 in. (64 x 64 x 56 cm)
Length (English)25 in.
Length (Metric)64 cm
Width (English)25 in.
Width (English) Interior14 in.
Width (Metric)64 cm
Width (Metric) Interior36 cm
Height (English)22 in.
Height (English) Interior14 in.
Height (Metric)56 cm
Height (Metric) Interior36 cm
Depth (English) Interior20 in.
Depth (Metric) Interior51 cm
Shipping Weight (English)295 lb.
Shipping Weight (Metric)134 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Wattage1600 w
Amperage6.7 A