Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Actual product may vary
Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Pick the optimal choice for a busy lab with these dedicated platforms, which are model-specific and come with pre-installed clamps to maximize capacity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeHoldsDimensions (L x W)For Use With (Equipment)
3523-49Adjustable Vessel Platform11 x 13 in. (28 x 33 cm)MaxQ™ 2000, 2506, 2508 Shaker
3523-46Shaker Platform Utility TrayTray11 x 13 in. (28 x 33 cm)MaxQ™ 2000, 2506, 2508, 4450 Shaker
Catalog number 3523-49
Price (SGD)
2,729.00
EA
Add to cart
Product Type:
Adjustable Vessel Platform
Dimensions (L x W):
11 x 13 in. (28 x 33 cm)
For Use With (Equipment):
MaxQ™ 2000, 2506, 2508 Shaker
Price (SGD)
2,729.00
EA
Add to cart
Pick the optimal choice for a busy lab with Thermo Scientific™ Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers. MaxQ platforms are model-specific and come with pre-installed clamps to maximize capacity. MaxQ shakers, platforms and accessories provide exceptional innovation, reliability and accuracy.
Clamps come installed to provide maximum capacity.
Specifications
Product TypeAdjustable Vessel Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)11 x 13 in. (28 x 33 cm)
Length (English) Platform11 in.
Length (Metric) Platform28 cm
Width (English) Platform13 in.
Width (Metric) Platform33 cm
DescriptionAdjustable vessel platform
For Use With (Equipment)MaxQ™ 2000, 2506, 2508 Shaker
Unit SizeEA