Search Fisher Scientific
| Catalog Number | Bottom Shape | Capacity (Metric) | Thread Style | Style |
|---|---|---|---|---|
| 351934 | Round | 1.8 mL | Internal | Writing Surface, No Cap |
| 374080 | Conical (Self-standing) | 0.5 mL | Internal | No Writing Surface, A-R Base |
| 375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
| FIS12-565-164N | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
| FKS10544641 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
| NNC375353 | Conical (Self-standing) | 1 mL | External | Writing Surface, A-R Base |
| FIS12-565-297 | Conical (Self-standing) | 1 mL | External | No Writing Surface, A-R Base |
| NNC377224 | Conical (Self-standing) | 1 mL | Internal | Writing Surface, A-R Base |
| 366656 | Conical (Self-standing) | 1 mL | Internal | Writing Surface |
| 375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base |
| NNC375418 | Round (Self-standing) | 1.8 mL | External | Writing Surface, A-R Base |
| 347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
| FKS10080601 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
| NNC347627 | Round (Self-standing) | 1.8 mL | External | No Writing Surface, A-R Base |
| 377267 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
| FIS12-565-167N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
| FKS11341675 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface, A-R Base |
| 368632 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
| FIS12-565-171N | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
| FKS10674511 | Round (Self-standing) | 1.8 mL | Internal | Writing Surface |
| 363401 | Round | 1.8 mL | Internal | Writing Surface |
| FKS10004220 | Round | 1.8 mL | Internal | Writing Surface |
| NNC363401 | Round | 1.8 mL | Internal | Writing Surface |
| 379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
| FIS12-565-162N | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
| NNC379189 | Round (Self-standing) | 3.6 mL | Internal | Writing Surface, A-R Base |
| FKS10789871 | Round | 3.6 mL | Internal | Writing Surface |
| 337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
| FIS12-565-291 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
| NNC337516 | Round (Self-standing) | 4.5 mL | External | Writing Surface, A-R Base |
| 347643 | Round (Self-standing) | 4.5 mL | External | No Writing Surface, A-R Base |
| FIS12-565-161N | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base |
| FKS10716922 | Conical (Self-standing) | 4.5 mL | Internal | Writing Surface, A-R Base |
| 363452 | Round | 4.5 mL | Internal | Writing Surface |
| FIS12-565-173N | Round | 4.5 mL | Internal | Writing Surface |
| FKS10358131 | Round | 4.5 mL | Internal | Writing Surface |
Features: