Hands-Free Fold-Up Magnifiers
Hands-Free Fold-Up Magnifiers
Actual product may vary
Hands-Free Fold-Up Magnifiers
Allow two-handed examination of objects while yielding a sharp, crisp and distinct image
Have Questions?
Catalog number 3434
also known as 3434V
Price (SGD)
36.00
PK
Add to cart
Price (SGD)
36.00
PK
Add to cart
Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
  • Bottom panel features white grid markings in both inches and millimeters to assist in measuring
  • All-glass optics ensure perfect focus with zero distortion
  • Seven sizes are available, including three featuring brilliant battery-powered illumination
Specifications
Cap StyleFlat
For Use With (Application)For use in quality control, labs, food, electronics, manufacturing and universities
Diameter (Englis) Lens1.25 in.
Diameter (Metric) Lens3.17 cm
Magnification5x
MaterialGlass, Glass
Product TypeMagnifier (Hands-free, Fold-up)
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Capacity (Metric)2 mL
ColorNatural
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
SterilityNon-sterile
GraduatedYes
Max. RCF25,000 x g
Unit SizePK